Gene Transcripts

Gene Symbol
Prss54
Entrez Gene ID
70993
Gene Name
protease, serine 54
Gene Aliases
4931432M23Rik, Klkbl4
Chromosome Location
Chr. 8: 95559067 - 95576330 on Build GRCm38
UniGene ID
Mm.486561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027640.1 NP_081916.1 2 96
GenBank mRNA AK016501.1 - 4 447
BC144859.1 - 3 223 9053407
BC145787.1 - 3 223 40131090

View Full Product Details