Gene Transcripts

Gene Symbol
Wdsub1
Entrez Gene ID
72137
Gene Name
WD repeat, SAM and U-box domain containing 1
Gene Aliases
1700048E19Rik, 2610014F08Rik
Chromosome Location
Chr. 2: 59852364 - 59882625 on Build GRCm38
UniGene ID
Mm.288734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159636.1 NP_001153108.1 7, 8 1069
NM_028118.2 NP_082394.1 7, 8 1069
GenBank mRNA AK011391.1 - 7, 8 1005
BC050792.1 - 7, 8 1010
BC061948.1 - 7, 8 1047 6312489

View Full Product Details