Gene Transcripts
- Gene Symbol
- Tonsl
- Entrez Gene ID
- 72749
- Gene Name
- tonsoku-like, DNA repair protein
- Gene Aliases
- 2810439M11Rik, Nfkbil2
- Chromosome Location
- Chr. 15: 76626237 - 76639929 on Build GRCm38
- UniGene ID
- Mm.358749
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_183091.3 |
NP_898914.3
|
18 |
2830 |
|
GenBank mRNA |
BC066068.1 |
- |
18 |
2812 |
|
View Full Product Details