Gene Transcripts

Gene Symbol
Veph1
Entrez Gene ID
72789
Gene Name
ventricular zone expressed PH domain-containing 1
Gene Aliases
2810471M23Rik, Veph
Chromosome Location
Chr. 3: 66053558 - 66297066 on Build GRCm38
UniGene ID
Mm.443526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145820.3 NP_665819.2 14 2763
GenBank mRNA AB085187.1 - 14 2745
AB085189.1 - 6 808
AK013398.1 - 6 823
BC075649.1 - 14 2761
BC094339.1 - 6 776
BC128271.1 - 13 2470 40059997

View Full Product Details