Gene Transcripts

Gene Symbol
Tssk5
Entrez Gene ID
73542
Gene Name
testis-specific serine kinase 5
Gene Aliases
1700091F14Rik
Chromosome Location
Chr. 15: 76371958 - 76375191 on Build GRCm38
UniGene ID
Mm.159026
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183099.2 NP_898922.2 4 805
GenBank mRNA AK007032.1 - 4 807
BC114543.1 - 4 609

View Full Product Details