Gene Transcripts

Gene Symbol
Palm3
Entrez Gene ID
74337
Gene Name
paralemmin 3
Gene Aliases
4432412L15Rik, AL024300
Chromosome Location
Chr. 8: 84021474 - 84030295 on Build GRCm38
UniGene ID
Mm.250943
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028877.1 NP_083153.1 7 1677
GenBank mRNA BC113196.1 - 7 1680
BC113802.1 - 7 1675
EF208034.1 - 7 1677

View Full Product Details