Gene Transcripts

Gene Symbol
Lonrf3
Entrez Gene ID
74365
Gene Name
LON peptidase N-terminal domain and ring finger 3
Gene Aliases
4932412G04Rik, 5730439E01Rik, A830039N02Rik, AU023707, Rnf127
Chromosome Location
Chr. X: 36328358 - 36366857 on Build GRCm38
UniGene ID
Mm.327654
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028894.1 NP_083170.1 7 1758
GenBank mRNA AK016522.1 - 7 1758
AK167341.1 - 7 1817
BC120688.1 - 7 1660 40129611
BC120690.1 - 7 1660 40129613

View Full Product Details