Gene Transcripts

Gene Symbol
Mettl5
Entrez Gene ID
75422
Gene Name
methyltransferase like 5
Gene Aliases
2810410A08Rik
Chromosome Location
Chr. 2: 69871195 - 69885984 on Build GRCm38
UniGene ID
Mm.10508
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029280.4 NP_083556.2 3 610
GenBank mRNA AK013063.1 - 3 593
AK133594.1 - 3 600
BC027547.1 - 3 543 1531973
CJ063020.1 - 3 600

View Full Product Details