Gene Transcripts

Gene Symbol
NDUFV3
Entrez Gene ID
4731
Gene Name
NADH:ubiquinone oxidoreductase subunit V3
Gene Aliases
CI-10k, CI-9KD
Chromosome Location
Chr. 21: 42893268 - 42911396 on Build GRCh38
UniGene ID
Hs.473937
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001503.1 NP_001001503.1 3 261
NM_021075.3 NP_066553.3 4 1356
XM_011529586.2 XP_011527888.1 5 1416
XM_017028359.1 XP_016883848.1 4 319
GenBank mRNA AK056041.1 - 6 1813
AK289586.1 - 3 215
AK308228.1 - 4 1313
AY173949.1 - 3 219
BC021217.2 - 4 1343 4302265
BC033766.1 - 3 219
BC054016.1 - 3 262
CR542174.1 - 3 195
CR542190.1 - 3 195

View Full Product Details