Gene Transcripts

Gene Symbol
Urah
Entrez Gene ID
76974
Gene Name
urate (5-hydroxyiso-) hydrolase
Gene Aliases
1190003J15Rik, 2810420C16Rik
Chromosome Location
Chr. 7: 140835023 - 140837968 on Build GRCm38
UniGene ID
Mm.272359
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029821.2 NP_084097.1 3 429
GenBank mRNA AK004470.1 - 3 647
AK013117.1 - 3 431
BC051545.1 - 3 597 5255010
DQ225768.1 - 3 304

View Full Product Details