Gene Transcripts

Gene Symbol
Armcx1
Entrez Gene ID
78248
Gene Name
armadillo repeat containing, X-linked 1
Gene Aliases
3010033I09Rik, ALEX1
Chromosome Location
Chr. X: 134717912 - 134721917 on Build GRCm38
UniGene ID
Mm.458522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166377.1 NP_001159849.1 6 1303
NM_001166378.1 NP_001159850.1 4 1002
NM_001166379.1 NP_001159851.1 4 1118
NM_001166380.1 NP_001159852.1 5 1094
NM_030066.3 NP_084342.1 6 1187
GenBank mRNA AK019405.1 - 6 1114
AK147002.1 - 6 1254
BC021410.1 - 5 1022 5007058
BC026488.1 - 4 1086 4504219
BC068228.1 - 6 1136 6413546

View Full Product Details