Gene Transcripts
- Gene Symbol
- Pus7
- Entrez Gene ID
- 78697
- Gene Name
- pseudouridylate synthase 7 homolog (S. cerevisiae)
- Gene Aliases
- C330017I15Rik
- Chromosome Location
- Chr. 5: 23740165 - 23783711 on Build GRCm38
- UniGene ID
- Mm.366825
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289780.1 |
NP_001276709.1
|
15, 16 |
2012 |
|
NM_001289781.1 |
NP_001276710.1
|
14, 15 |
2345 |
|
NM_178403.5 |
NP_848490.2
|
14, 15 |
1994 |
|
GenBank mRNA |
AK142271.1 |
- |
14, 15 |
1963 |
|
AK145380.1 |
- |
14, 15 |
2345 |
|
AK160849.1 |
- |
14, 15 |
1946 |
|
AK173290.1 |
- |
14, 15 |
1934 |
|
BC008544.1 |
- |
14, 15 |
1948 |
3497634 |
BC010600.1 |
- |
6, 7 |
729 |
|
BC138533.1 |
- |
14, 15 |
1836 |
8861553 |
BC145316.1 |
- |
15, 16 |
1854 |
9053871 |
View Full Product Details