Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001082476.2 | NP_001075945.1 | 3 | 292 | |
NM_001252541.1 | NP_001239470.1 | 3 | 292 | ||
NM_001252542.1 | NP_001239471.1 | 3 | 359 | ||
GenBank mRNA | AK019617.1 | - | 3 | 289 | |
AK143124.1 | - | 3 | 332 | ||
AK163207.1 | - | 3 | 262 | ||
AK169885.1 | - | 3 | 351 | ||
BC049789.1 | - | 3 | 342 | 6331033 | |
BQ771207.1 | - | 3 | 259 | ||
CD354207.1 | - | 683 |