Gene Transcripts

Gene Symbol
Clmn
Entrez Gene ID
94040
Gene Name
calmin
Gene Aliases
9330188N17Rik, AI428889, AI788815, mKIAA1188
Chromosome Location
Chr. 12: 104763114 - 104865076 on Build GRCm38
UniGene ID
Mm.244078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040682.1 NP_001035772.1 9 2347
NM_053155.2 NP_444385.2 9 2347
GenBank mRNA AB047978.1 - 9 2340
AB059643.1 - 9 2340
AB059644.1 - 9 2340
AB059645.1 - 9 2340
AB059646.1 - 9 2340
AB059647.1 - 9 2340
AB059648.1 - 9 2340
AK077023.1 - 6 1929
AK147777.1 - 9 2244
AK162015.1 - 1 2064
AK220261.1 - 1 326
BC019176.1 - 9 2278
BC096470.1 - 9 2236
BC141280.1 - 9 2280 9055892

View Full Product Details