Gene Transcripts

Gene Symbol
Mael
Entrez Gene ID
98558
Gene Name
maelstrom homolog (Drosophila)
Gene Aliases
4933405K18Rik, AU019877
Chromosome Location
Chr. 1: 166201385 - 166238744 on Build GRCm38
UniGene ID
Mm.26189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175296.4 NP_780505.1 11 1163
GenBank mRNA AK077103.1 - 11 1160
AK165801.1 - 11 1160
BC050800.1 - 11 1163 6744032

View Full Product Details