Gene Transcripts

Gene Symbol
Xpo1
Entrez Gene ID
103573
Gene Name
exportin 1, CRM1 homolog (yeast)
Gene Aliases
AA420417, Crm1, Exp1
Chromosome Location
Chr. 11: 23255914 - 23297597 on Build GRCm38
UniGene ID
Mm.217547
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001035226.1 NP_001030303.1 16 1820
NM_134014.3 NP_598775.2 16 2058
GenBank mRNA AK136538.1 - 9 1221
BC012276.1 - 3 282 3156478
BC025628.1 - 2 242 5321933
BC029714.1 - 14 1581
BC034710.1 - 15 1815
BC062912.1 - 16 1820 6821729
BU052960.1 - 2 163
CF538023.1 - 5 661

View Full Product Details