Gene Transcripts

Gene Symbol
Unc5a
Entrez Gene ID
107448
Gene Name
unc-5 homolog A (C. elegans)
Gene Aliases
Unc5h1, mKIAA1976
Chromosome Location
Chr. 13: 54949417 - 55006018 on Build GRCm38
UniGene ID
Mm.23573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153131.3 NP_694771.1 3 552
GenBank mRNA AJ487852.1 - 3 553
AK122575.1 - 3 320
BC058084.1 - 3 573 6813463

View Full Product Details