Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029770.2 | NP_084046.2 | 9 | 1607 | |
GenBank mRNA | AJ487853.1 | - | 9 | 1510 | |
AK147603.1 | - | 8 | 1469 | ||
AK154271.1 | - | 9 | 1607 | ||
AK154287.1 | - | 9 | 1610 | ||
AK158284.1 | - | 9 | 1607 | ||
AK170513.1 | - | 9 | 1606 | ||
BC048162.1 | - | 8 | 1381 | ||
BC057560.1 | - | 8 | 1381 | 6417563 |