Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145537.2 | NP_663512.2 | 9 | 1147 | |
GenBank mRNA | AK029164.1 | - | 9 | 1099 | |
AK075650.1 | - | 9 | 1061 | ||
AK079296.1 | - | 9 | 1083 | ||
AK080636.1 | - | 9 | 1092 | ||
AK149892.1 | - | 9 | 1082 | ||
AK155024.1 | - | 9 | 1110 | ||
AK166998.1 | - | 9 | 1083 | ||
BC008268.1 | - | 9 | 1058 | 2811074 |