Gene Transcripts

Gene Symbol
ACYP2
Entrez Gene ID
98
Gene Name
acylphosphatase 2
Gene Aliases
ACYM, ACYP
Chromosome Location
Chr. 2: 53970838 - 54305300 on Build GRCh38
UniGene ID
Hs.516173
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320586.1 NP_001307515.1 6 599
NM_001320587.1 NP_001307516.1 5 506
NM_001320588.1 NP_001307517.1 3 300
NM_001320589.1 NP_001307518.1 3 551
NM_001320590.1 NP_001307519.1 3 551
NM_138448.3 NP_612457.1 3 551
XM_017005406.1 XP_016860895.1 7 945
XM_017005407.1 XP_016860896.1 6 864
XM_017005408.1 XP_016860897.1 5 1573
XM_017005409.1 XP_016860898.1 5 771
XM_017005410.1 XP_016860899.1 3 421
XM_017005411.1 XP_016860900.1 7 945
XM_017005412.1 XP_016860901.1 7 945
XM_017005413.1 XP_016860902.1 7 945
GenBank mRNA AK307096.1 - 3 422
BC012290.1 - 3 320 3864319
BF671963.1 - 3 299
BG741807.1 - 5 500
BU543943.1 - 6 599
BU857820.1 - 3 409

View Full Product Details