Gene Transcripts

Gene Symbol
Xylt1
Entrez Gene ID
233781
Gene Name
xylosyltransferase 1
Gene Aliases
8030490L12
Chromosome Location
Chr. 7: 117380979 - 117667630 on Build GRCm38
UniGene ID
Mm.461427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175645.3 NP_783576.2 2, 3 589
GenBank mRNA AJ539164.1 - 2, 3 591
AK033309.1 - 2, 3 591
EF410103.1 - 301

View Full Product Details