Gene Transcripts

Gene Symbol
Parp16
Entrez Gene ID
214424
Gene Name
poly (ADP-ribose) polymerase family, member 16
Gene Aliases
ARTD15, BC055447, C79952, PARP-16
Chromosome Location
Chr. 9: 65214380 - 65239220 on Build GRCm38
UniGene ID
Mm.31129
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177460.4 NP_803411.3 3, 4 598
GenBank mRNA AK033791.1 - 548
AK133475.1 - 3, 4 599
AK166002.1 - 2, 3 362
AK169866.1 - 3, 4 513
BC014731.1 - 2, 3 628
BC055447.1 - 3, 4 567 3594406

View Full Product Details