Gene Transcripts

Gene Symbol
SCYL2
Entrez Gene ID
55681
Gene Name
SCY1 like pseudokinase 2
Gene Aliases
CVAK104
Chromosome Location
Chr. 12: 100267137 - 100341724 on Build GRCh38
UniGene ID
Hs.506481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317784.1 NP_001304713.1 9, 10 1553
NM_017988.5 NP_060458.3 9, 10 1705
GenBank mRNA AK000936.1 - 8, 9 1311
AK024274.1 - 5, 6 714
AK027551.1 - 5, 6 771
AK125091.1 - 9, 10 1690
AK292980.1 - 9, 10 1479
BC063798.1 - 9, 10 1318 6473586

View Full Product Details