Gene Transcripts

Gene Symbol
KCNJ14
Entrez Gene ID
3770
Gene Name
potassium voltage-gated channel subfamily J member 14
Gene Aliases
IRK4, KIR2.4
Chromosome Location
Chr. 19: 48455707 - 48466110 on Build GRCh38
UniGene ID
Hs.144011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013348.3 NP_037480.1 3 1847
GenBank mRNA AF081466.1 - 3 2045
BC035918.1 - 2 2135 5531571

View Full Product Details