Gene Transcripts
- Gene Symbol
- CAMKV
- Entrez Gene ID
- 79012
- Gene Name
- CaM kinase like vesicle associated
- Gene Aliases
- 1G5, VACAMKL
- Chromosome Location
- Chr. 3: 49857981 - 49870222 on Build GRCh38
- UniGene ID
- Hs.145156
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320147.1 |
NP_001307076.1
|
2, 3 |
571 |
|
NM_024046.4 |
NP_076951.2
|
2, 3 |
571 |
|
GenBank mRNA |
AK074856.1 |
- |
2, 3 |
285 |
|
AK074899.1 |
- |
2, 3 |
255 |
|
AK075294.1 |
- |
2, 3 |
266 |
|
AK295588.1 |
- |
2, 3 |
258 |
|
AK297437.1 |
- |
2, 3 |
252 |
|
AK299953.1 |
- |
2, 3 |
254 |
|
AK315525.1 |
- |
2, 3 |
304 |
|
AL136697.1 |
- |
2, 3 |
254 |
|
AL833943.1 |
- |
2, 3 |
255 |
|
BC000497.2 |
- |
2, 3 |
240 |
2820734 |
BC001921.1 |
- |
2, 3 |
240 |
2820734 |
BC005828.2 |
- |
2, 3 |
240 |
2820734 |
BC017363.2 |
- |
2, 3 |
242 |
3940018 |
BC019256.1 |
- |
2, 3 |
235 |
3502894 |
BX647074.1 |
- |
2, 3 |
252 |
|
CR533508.1 |
- |
1, 2 |
92 |
|
DC351900.1 |
- |
|
571 |
|
View Full Product Details