Gene Transcripts

Gene Symbol
ROBO1
Entrez Gene ID
6091
Gene Name
roundabout guidance receptor 1
Gene Aliases
DUTT1, SAX3
Chromosome Location
Chr. 3: 78597238 - 79767998 on Build GRCh38
UniGene ID
Hs.744218
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145845.1 NP_001139317.1 10 2368
NM_002941.3 NP_002932.1 11 1686
NM_133631.3 NP_598334.2 10 2368
XM_006713277.2 XP_006713340.1 10 2337
XM_011533976.1 XP_011532278.1 12 1784
XM_011533977.2 XP_011532279.1 12 1637
XM_011533978.1 XP_011532280.1 12 1784
XM_011533979.1 XP_011532281.1 12 1784
XM_011533980.1 XP_011532282.1 12 1784
XM_017006982.1 XP_016862471.1 11 1696
XM_017006983.1 XP_016862472.1 9 2328
XM_017006984.1 XP_016862473.1 11 1775
XM_017006985.1 XP_016862474.1 10 1437
GenBank mRNA AF040990.1 - 10 1478
AK095256.1 - 5 1303
AK299920.1 - 1 375
BC112336.1 - 10 1421 40037251
BC115020.1 - 10 1469 7961997
BC115022.1 - 10 1469 7962000
BC157861.1 - 10 1665 9057080
BC171855.1 - 10 1665 9054509
BX538319.1 - 3 385

View Full Product Details