Gene Transcripts
- Gene Symbol
- SLC27A4
- Entrez Gene ID
- 10999
- Gene Name
- solute carrier family 27 member 4
- Gene Aliases
- ACSVL4, FATP4, IPS
- Chromosome Location
- Chr. 9: 128340517 - 128361470 on Build GRCh38
- UniGene ID
- Hs.656699
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005094.3 |
NP_005085.2
|
10, 11 |
1742 |
|
XM_017014222.1 |
XP_016869711.1
|
11, 12 |
1615 |
|
GenBank mRNA |
AF055899.1 |
- |
9, 10 |
1454 |
|
AK000722.1 |
- |
1, 2 |
104 |
|
AK027504.1 |
- |
9, 10 |
1322 |
|
AK074831.1 |
- |
7, 8 |
790 |
|
AK290222.1 |
- |
10, 11 |
1742 |
|
AK314681.1 |
- |
9, 10 |
1762 |
|
BC004268.1 |
- |
3, 4 |
302 |
|
BC009959.2 |
- |
2, 4 |
355 |
|
BC065003.1 |
- |
10, 11 |
1741 |
6023438 |
View Full Product Details