Gene Transcripts
- Gene Symbol
- GRHL3
- Entrez Gene ID
- 57822
- Gene Name
- grainyhead like transcription factor 3
- Gene Aliases
- SOM, TFCP2L4, VWS2
- Chromosome Location
- Chr. 1: 24319322 - 24364482 on Build GRCh38
- UniGene ID
- Hs.657920
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195010.1 |
NP_001181939.1
|
3, 4 |
439 |
|
NM_021180.3 |
NP_067003.2
|
3, 4 |
335 |
|
NM_198173.2 |
NP_937816.1
|
3, 4 |
504 |
|
NM_198174.2 |
NP_937817.3
|
3, 4 |
504 |
|
XM_011541869.1 |
XP_011540171.1
|
3, 4 |
385 |
|
XM_011541870.2 |
XP_011540172.1
|
2, 3 |
306 |
|
GenBank mRNA |
AK074386.1 |
- |
3, 4 |
401 |
|
AK315164.1 |
- |
3, 4 |
306 |
|
AY231160.1 |
- |
3, 4 |
335 |
|
AY231161.1 |
- |
2, 3 |
148 |
|
BC036890.2 |
- |
3, 4 |
450 |
5502632 |
BU848928.1 |
- |
3, 4 |
435 |
|
DA714132.1 |
- |
3, 4 |
504 |
|
View Full Product Details