Gene Transcripts

Gene Symbol
OIT3
Entrez Gene ID
170392
Gene Name
oncoprotein induced transcript 3
Gene Aliases
LZP
Chromosome Location
Chr. 10: 72893556 - 72933036 on Build GRCh38
UniGene ID
Hs.8366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152635.2 NP_689848.1 2 630
NR_130125.1 2 630
GenBank mRNA AK096435.1 - 2 605
AY013707.2 - 2 461
AY358339.1 - 2 583
BC126440.1 - 2 602 8992156

View Full Product Details