Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001359.1 | NP_001350.1 | 2 | 297 | |
XM_011516880.2 | XP_011515182.1 | 3 | 292 | ||
XM_017013147.1 | XP_016868636.1 | 4 | 952 | ||
XM_017013148.1 | XP_016868637.1 | 4 | 692 | ||
GenBank mRNA | AK300069.1 | - | 4 | 812 | |
BC105080.1 | - | 2 | 244 | 8144083 | |
BC105082.1 | - | 2 | 244 | 8144085 | |
L26050.1 | - | 2 | 166 | ||
U49352.1 | - | 2 | 195 |