Gene Transcripts

Gene Symbol
PRSS22
Entrez Gene ID
64063
Gene Name
protease, serine 22
Gene Aliases
BSSP-4, SP001LA, hBSSP-4
Chromosome Location
Chr. 16: 2852727 - 2858630 on Build GRCh38
UniGene ID
Hs.459709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022119.3 NP_071402.1 4 359
XM_005255473.3 XP_005255530.1 5 458
GenBank mRNA AB010779.1 - 4 296
AF321182.1 - 4 310
AK309624.1 - 4 501
AY358396.1 - 4 345
BC009726.1 - 4 331 3899480
BQ893064.1 - 367

View Full Product Details