Gene Transcripts

Gene Symbol
CYP4X1
Entrez Gene ID
260293
Gene Name
cytochrome P450 family 4 subfamily X member 1
Gene Aliases
CYPIVX1
Chromosome Location
Chr. 1: 46961364 - 47055432 on Build GRCh38
UniGene ID
Hs.439760
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320289.1 NP_001307218.1 5 754
NM_001320290.1 NP_001307219.1 5 491
NM_178033.1 NP_828847.1 5 788
XM_017000973.1 XP_016856462.1 5 819
GenBank mRNA AK091806.1 - 5 492
AK098065.1 - 5 781
AK131355.1 - 5 754
AM040940.1 - 5 625
AY358537.1 - 5 771
BC028102.1 - 5 788 5243005

View Full Product Details