Gene Transcripts

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
Chromosome Location
Chr. 16: 3725054 - 3880727 on Build GRCh38
UniGene ID
Hs.459759
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079846.1 NP_001073315.1 2 461
NM_004380.2 NP_004371.2 2 461
XM_005255124.4 XP_005255181.1 2 1068
XM_005255125.4 XP_005255182.1 2 1068
XM_006720848.3 XP_006720911.1 2 1068
XM_011522382.2 XP_011520684.1 2 1068
XM_017022944.1 XP_016878433.1 2 1068
GenBank mRNA AB210043.1 - 2 461
U47741.1 - 3 455
U85962.3 - 2 1075

View Full Product Details