Gene Transcripts

Gene Symbol
DAO
Entrez Gene ID
1610
Gene Name
D-amino acid oxidase
Gene Aliases
DAAO, DAMOX, OXDA
Chromosome Location
Chr. 12: 108880073 - 108901043 on Build GRCh38
UniGene ID
Hs.113227
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001917.4 NP_001908.3 3 364
XM_005268692.3 XP_005268749.1 3 364
XM_011538004.2 XP_011536306.1 4 450
XM_011538005.2 XP_011536307.1 4 454
GenBank mRNA AK312995.1 - 3 351
AX741655.1 - 3 354
AX741656.1 - 3 354
AX741659.1 - 3 324
AX776478.1 - 3 354
AX776479.1 - 3 354
AX776480.1 - 6 1366
AX786386.1 - 3 354
BC029057.1 - 3 334 5184628
BC074770.2 - 3 236 30915193
DA085245.1 - 3 365
KU178015.1 - 2 211
KU178016.1 - 2 211

View Full Product Details