Gene Transcripts

Gene Symbol
CMYA5
Entrez Gene ID
202333
Gene Name
cardiomyopathy associated 5
Gene Aliases
C5orf10, SPRYD2, TRIM76
Chromosome Location
Chr. 5: 79689339 - 79800226 on Build GRCh38
UniGene ID
Hs.482625
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153610.4 NP_705838.3 2 8876
XM_017009212.1 XP_016864701.1 2 8744
GenBank mRNA AK123466.1 - 1 234
AL832021.1 - 1 1250
AL832368.1 - 1 483
AL834252.2 - 1 593
BC046215.1 - 1 1513
BC062664.1 - 1 1030
BC111529.2 - 1 186
BC111530.2 - 1 186
BX647452.1 - 1 5022
BX647628.1 - 1 580
BX647646.1 - 1 580

View Full Product Details