Gene Transcripts

Gene Symbol
SLC5A4
Entrez Gene ID
6527
Gene Name
solute carrier family 5 member 4
Gene Aliases
DJ90G24.4, SAAT1, SGLT3
Chromosome Location
Chr. 22: 32218423 - 32355057 on Build GRCh38
UniGene ID
Hs.130101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014227.2 NP_055042.1 14 1717
XM_006724308.3 XP_006724371.1 15 1968
XM_011530342.2 XP_011528644.1 16 2086
XM_011530343.2 XP_011528645.1 14 1632
XM_011530344.2 XP_011528646.1 13 1597
XM_017028920.1 XP_016884409.1 16 2047
GenBank mRNA AJ133127.1 - 14 1731
GQ292716.1 - 3 445
JQ340171.1 - 3 445
JQ519903.1 - 3 445

View Full Product Details