Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007126.3 | NP_009057.1 | 17 | 3014 | |
GenBank mRNA | AL137377.1 | - | 8 | 1500 | |
BC007562.1 | - | 13 | 2140 | ||
BC110913.1 | - | 17 | 2824 | 6502535 | |
BC121794.1 | - | 17 | 2821 | 40108360 | |
BC122550.1 | - | 16 | 2899 | 40108357 | |
FJ224344.1 | - | 17 | 2836 |