Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040427.1 | NP_001035517.1 | 8 | 1174 | |
NM_001286249.1 | NP_001273178.1 | 6 | 969 | ||
NM_005326.4 | NP_005317.2 | 7 | 1078 | ||
XM_011522469.2 | XP_011520771.1 | 7 | 718 | ||
XM_011522470.2 | XP_011520772.1 | 6 | 607 | ||
XM_017023195.1 | XP_016878684.1 | 7 | 643 | ||
GenBank mRNA | AK290155.1 | - | 7 | 643 | |
AK298174.1 | - | 6 | 609 | ||
AK299173.1 | - | 7 | 709 | ||
AK299993.1 | - | 7 | 709 | ||
BC000840.1 | - | 7 | 698 | 3457214 | |
BC002627.2 | - | 7 | 1027 | 3608098 | |
X90999.1 | - | 6 | 563 |