Gene Transcripts

Gene Symbol
MORC1
Entrez Gene ID
27136
Gene Name
MORC family CW-type zinc finger 1
Gene Aliases
CT33, MORC, ZCW6
Chromosome Location
Chr. 3: 108958239 - 109118146 on Build GRCh38
UniGene ID
Hs.591296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014429.3 NP_055244.3 6, 7 518
XM_005247362.2 XP_005247419.1 6, 7 475
XM_011512691.1 XP_011510993.1 6, 7 475
XM_011512692.1 XP_011510994.1 6, 7 475
XM_011512693.1 XP_011510995.1 6, 7 475
XM_011512694.1 XP_011510996.1 3, 4 226
XM_017006169.1 XP_016861658.1 6, 7 475
GenBank mRNA AF084946.1 - 6, 7 493
AK302642.1 - 6, 7 475
BC028680.1 - 6, 7 519
BC050307.1 - 6, 7 457 4838254

View Full Product Details