Gene Transcripts

Gene Symbol
GUCA1B
Entrez Gene ID
2979
Gene Name
guanylate cyclase activator 1B
Gene Aliases
GCAP2, GUCA2, RP48
Chromosome Location
Chr. 6: 42183284 - 42194956 on Build GRCh38
UniGene ID
Hs.446529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002098.5 NP_002089.4 2 385
GenBank mRNA BQ636025.1 - 2 362
BX537393.1 - 2 385

View Full Product Details