Gene Transcripts

Gene Symbol
UBE2D1
Entrez Gene ID
7321
Gene Name
ubiquitin conjugating enzyme E2 D1
Gene Aliases
E2(17)KB1, SFT, UBC4/5, UBCH5, UBCH5A
Chromosome Location
Chr. 10: 58334979 - 58370753 on Build GRCh38
UniGene ID
Hs.129683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204880.1 NP_001191809.1 6 1774
NM_003338.4 NP_003329.1 7 1806
XM_017016609.1 XP_016872098.1 7 1705
GenBank mRNA BM468116.1 - 1 510

View Full Product Details