Gene Transcripts

Gene Symbol
ATP2C1
Entrez Gene ID
27032
Gene Name
ATPase secretory pathway Ca2+ transporting 1
Gene Aliases
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
Chromosome Location
Chr. 3: 130850500 - 131016712 on Build GRCh38
UniGene ID
Hs.584884
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001485.2 NP_001001485.1 18 1931
NM_001001486.1 NP_001001486.1 17 1608
NM_001001487.1 NP_001001487.1 17 1608
NM_001199179.1 NP_001186108.1 18 1837
NM_001199180.1 NP_001186109.1 17 1826
NM_001199181.1 NP_001186110.1 17 1826
NM_001199182.1 NP_001186111.1 16 1709
NM_001199183.1 NP_001186112.1 17 1699
NM_001199184.1 NP_001186113.1 17 1699
NM_001199185.1 NP_001186114.1 17 1750
NM_014382.3 NP_055197.2 17 1750
XM_005247354.2 XP_005247411.1 17 1851
XM_005247355.2 XP_005247412.1 18 1978
XM_005247356.2 XP_005247413.1 18 1731
XM_005247358.2 XP_005247415.1 17 1555
XM_011512686.2 XP_011510988.1 8 660
XM_017006164.1 XP_016861653.1 18 1639
GenBank mRNA AB037768.1 - 18 1838
AB209265.1 - 6 1598
AF181120.1 - 17 1663
AF181121.1 - 17 1663
AF189723.2 - 17 1582
AF225981.1 - 17 1582
AJ010953.1 - 3 160
AK001684.1 - 5 358
AK074692.1 - 15 1289
AK296470.1 - 17 1753
AK299945.1 - 16 1465
AK304175.1 - 17 1555
AK304374.1 - 17 1582
AK314342.1 - 17 1612
AY268374.1 - 17 1557
AY268375.1 - 17 1557
BC028139.1 - 17 1620 5215224

View Full Product Details