Gene Transcripts

Gene Symbol
AGA
Entrez Gene ID
175
Gene Name
aspartylglucosaminidase
Gene Aliases
AGU, ASRG, GA
Chromosome Location
Chr. 4: 177430774 - 177442503 on Build GRCh38
UniGene ID
Hs.207776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000027.3 NP_000018.2 5 677
NM_001171988.1 NP_001165459.1 5 677
NR_033655.1 5 677
GenBank mRNA AK303370.1 - 4 470
AK312982.1 - 5 611
BC012392.1 - 5 600 3859317
CR541715.1 - 5 549
CX869892.1 - 5 554
M64073.1 - 5 552
M64075.1 - 3 245
M64076.1 - 3 245
X55762.1 - 5 576

View Full Product Details