Gene Transcripts

Gene Symbol
CCDC87
Entrez Gene ID
55231
Gene Name
coiled-coil domain containing 87
Gene Aliases
Chromosome Location
Chr. 11: 66590169 - 66593083 on Build GRCh38
UniGene ID
Hs.121072
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018219.2 NP_060689.2 1 826
GenBank mRNA AK001648.1 - 1 806
BC034469.2 - 1 826 4823402

View Full Product Details