Gene Transcripts

Gene Symbol
NKAIN1
Entrez Gene ID
79570
Gene Name
Na+/K+ transporting ATPase interacting 1
Gene Aliases
FAM77C
Chromosome Location
Chr. 1: 31179270 - 31239887 on Build GRCh38
UniGene ID
Hs.470259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024522.2 NP_078798.2 7 2897
XM_017002320.1 XP_016857809.1 6 2491
GenBank mRNA AK022712.1 - 6 2469
AL832547.1 - 4 2162

View Full Product Details