Gene Transcripts

Gene Symbol
MED10
Entrez Gene ID
84246
Gene Name
mediator complex subunit 10
Gene Aliases
L6, NUT2, TRG20
Chromosome Location
Chr. 5: 6371926 - 6378526 on Build GRCh38
UniGene ID
Hs.13885
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032286.2 NP_115662.2 4 859
GenBank mRNA AK075493.1 - 1 1673

View Full Product Details