Gene Transcripts

Gene Symbol
MMGT1
Entrez Gene ID
93380
Gene Name
membrane magnesium transporter 1
Gene Aliases
EMC5, TMEM32
Chromosome Location
Chr. X: 135962072 - 135973975 on Build GRCh38
UniGene ID
Hs.110702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173470.1 NP_775741.1 4 1959
GenBank mRNA AL157477.1 - 4 1959
BC033588.1 - 4 1959 5213486

View Full Product Details