Gene Transcripts

Gene Symbol
YIPF6
Entrez Gene ID
286451
Gene Name
Yip1 domain family member 6
Gene Aliases
FinGER6
Chromosome Location
Chr. X: 68498374 - 68572413 on Build GRCh38
UniGene ID
Hs.82719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173834.3 NP_776195.2 2 409
XM_011530923.2 XP_011529225.1 2 817
XM_011530924.1 XP_011529226.1 2 191
XM_017029450.1 XP_016884939.1 2 191
GenBank mRNA AK095555.1 - 2 191
AK292835.1 - 2 151
BC012469.1 - 2 147 4452699
CN311465.1 - 2 409

View Full Product Details