Gene Transcripts

Gene Symbol
SPATA12
Entrez Gene ID
353324
Gene Name
spermatogenesis associated 12
Gene Aliases
SRG5
Chromosome Location
Chr. 3: 57060132 - 57076068 on Build GRCh38
UniGene ID
Hs.129794
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181727.1 NP_859078.1 2 1853
XM_011533676.2 XP_011531978.1 4 2399
GenBank mRNA AY221117.1 - 2 1853
BC033337.1 - 2 1628 4838903

View Full Product Details