Gene Transcripts

Gene Symbol
CYYR1
Entrez Gene ID
116159
Gene Name
cysteine and tyrosine rich 1
Gene Aliases
C21orf95
Chromosome Location
Chr. 21: 26466209 - 26573404 on Build GRCh38
UniGene ID
Hs.37445
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320768.1 NP_001307697.1 4 829
NM_052954.3 NP_443186.2 4 826
NR_135472.1 5 996
GenBank mRNA AF401639.1 - 4 431
AK054581.1 - 4 706
AK223576.1 - 4 546
AK304124.1 - 5 876
AK313231.1 - 4 612
AL833200.1 - 4 637
AY061853.1 - 4 371
BC036761.1 - 4 614 5588436
JF693914.1 - 3 299
KC843341.1 - 3 266
KF154022.1 - 5 598

View Full Product Details